Learn More
Aviva Systems Biology Corp KCNIP2 Antibody - C-terminal region (ARP35489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNIP2. It was validated on Western Blot. . Alternate names: DKFZp566L1246, KCHIP2, MGC17241ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-464