Learn More
Aviva Systems Biology Corp KCNIP4 Antibody - N-terminal region (ARP34622_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34622_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNIP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNIP4. Alternate names: CALP, KCHIP4ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55486