Learn More
Aviva Systems Biology Corp KCNIP4 Antibody - N-terminal region (ARP34622_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34622_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNIP4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KCNIP4. Alternate names: CALP, KCHIP4ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55485