Learn More
Aviva Systems Biology Corp KCNJ1 Antibody - middle region (ARP35432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNJ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNJ1. Alternate names: KIR1.1, ROMK, ROMK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55731