Learn More
Aviva Systems Biology Corp KCNJ12 Antibody - middle region (ARP35315_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35315_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNJ12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNJ12. Alternate names: FLJ14167, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x, IRK-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55695