Learn More
Aviva Systems Biology Corp Kcnj3 Antibody - C-terminal region (ARP35081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kcnj3. It was validated on Western Blot. . Alternate names: GIRK1, Kcnf3, Kir3.1, MGC124233, MGC124234ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-442