Learn More
Aviva Systems Biology Corp KCNJ4 Antibody - middle region (ARP35167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNJ4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNJ4. Alternate names: HIR, HIRK2, HRK1, IRK3, Kir2.3, MGC142066, MGC142068, IRK-3ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55657