Learn More
Aviva Systems Biology Corp KCNJ9 Antibody - middle region (ARP35171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNJ9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNJ9. Alternate names: GIRK3, KIR3.3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55659