Learn More
Aviva Systems Biology Corp KCNK10 Antibody - N-terminal region (ARP35399_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35399_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNK10. Alternate names: TREK2, TREK-2, K2p10.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55724