Learn More
Aviva Systems Biology Corp KCNK13 Antibody - C-terminal region (ARP35347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK13. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KCNK13. Alternate names: THIK1, THIK-1, K2p13.1ApplicationIHC, WB. Species reactivity: Horse, Human, Pig, Rat
Catalog No. 50-014-53002