Learn More
Aviva Systems Biology Corp KCNK15 Antibody - middle region (ARP35350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNK15. Alternate names: K2p15.1, KCNK11, KCNK14, KIAA0237, KT3.3, TASK-5, TASK5, dJ781B1.1ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-55708