Learn More
Aviva Systems Biology Corp KCNK3 Antibody - C-terminal region (ARP35090_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35090_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KCNK3. Alternate names: TASK, TBAK1, K2p3.1, TASK-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-52986