Learn More
Aviva Systems Biology Corp KCNK9 Antibody - N-terminal region (ARP35260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNK9. Alternate names: K2p9.1, KT3.2, MGC138268, MGC138270, TASK-3, TASK3ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52997