Learn More
Aviva Systems Biology Corp KCNMA1 Antibody - middle region (ARP37677_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37677_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNMA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNMA1. Alternate names: BKTM, DKFZp686K1437, KCa1.1, MGC71881, MaxiK, SAKCA, SLO, SLO-ALPHA, mSLO1, SLO1, bA205K10.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56391