Learn More
Aviva Systems Biology Corp KCNMB3 Antibody - middle region (ARP35444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNMB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNMB3. Alternate names: KCNMB2, KCNMBL, HBETA3, BKBETA3, SLOBETA3ApplicationWB. Species reactivity: Dog, Human, Rat
Catalog No. 50-014-55736