Learn More
Aviva Systems Biology Corp KCNMB4 Antibody - middle region (ARP35241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNMB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNMB4. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55677