Learn More
Aviva Systems Biology Corp KCNN1 Antibody - C-terminal region (ARP35095_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35095_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KCNN1. Alternate names: SK1, hSK1, SKCA1, KCa2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-52989