Learn More
Aviva Systems Biology Corp KCNN2 Antibody - middle region (ARP35439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNN2. Alternate names: KCa2.2, SK2, SKCA2, hSK2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55734