Learn More
Aviva Systems Biology Corp KCNN3 Antibody - C-terminal region (ARP35097_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35097_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KCNN3. Alternate names: SK3, hSK3, SKCA3, KCa2.3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55643