Learn More
Aviva Systems Biology Corp KCNN4 Antibody - C-terminal region (ARP35098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KCNN4. Alternate names: IK1, IKCA1, KCA4, KCa3.1, SK4, hIKCa1, hKCa4, hSK4ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat
Catalog No. 50-014-52990