Learn More
Aviva Systems Biology Corp KCNQ1 Antibody - N-terminal region (ARP35529_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35529_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNQ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNQ1. Alternate names: LQT, RWS, WRS, LQT1, SQT2, ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, Kv1.9, Kv7.1, KVLQT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55773