Learn More
Aviva Systems Biology Corp KCNQ1 Antibody - N-terminal region (ARP34924_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34924_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNQ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNQ1. Alternate names: ATFB1, FLJ26167, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS, ATFB3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55609