Learn More
Aviva Systems Biology Corp KCNQ2 Antibody - middle region (ARP35459_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35459_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNQ2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human KCNQ2. Alternate names: EBN, BFNC, EBN1, ENB1, BFNS1, EIEE7, HNSPC, KV7.2, KCNA11, KVEBN1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55744