Learn More
Aviva Systems Biology Corp KCNQ2 Antibody - N-terminal region (ARP35456_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35456_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNQ2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNQ2. Alternate names: BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KV7.2, KVEBN1, BFNS1, EIEE7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55741