Learn More
Aviva Systems Biology Corp Kcnq2 Antibody - middle region (ARP35140_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35140_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kcnq2. It was validated on Western Blot. . Alternate names: HNSPC, KQT2, Nmf134ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-445