Learn More
Aviva Systems Biology Corp KCNQ4 Antibody - middle region (ARP35148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNQ4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNQ4. Alternate names: DFNA2, KV7.4, DFNA2AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53918