Learn More
Aviva Systems Biology Corp KCNRG Antibody - middle region (ARP35502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNRG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNRG. Alternate names: DLTETApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55762