Learn More
Aviva Systems Biology Corp KCNS1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP35101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNS1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KCNS1. Alternate names: KV9.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-443