Learn More
Aviva Systems Biology Corp Kcnt1 Antibody - C-terminal region (ARP49576_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49576_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kcnt1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C030030G16Rik, Slack, mKIAA1422, slo2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-437