Learn More
Aviva Systems Biology Corp KCTD10 Antibody - N-terminal region (ARP35370_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35370_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD10. Alternate names: ULRO61, MSTP028ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55714