Learn More
Aviva Systems Biology Corp KCTD11 Antibody - N-terminal region (ARP36786_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36786_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD11. Alternate names: REN, KCASH1, C17orf36, REN/KCTD11ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-56275