Learn More
Aviva Systems Biology Corp KCTD11 Antibody - N-terminal region (ARP35623_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35623_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD11. Alternate names: C17orf36, MGC129844, REN, REN/KCTD11, KCASH1ApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55792