Learn More
Aviva Systems Biology Corp KCTD13 Antibody - N-terminal region (ARP35522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD13. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD13. Alternate names: PDIP1, FKSG86, POLDIP1, hBACURD1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55771