Learn More
Aviva Systems Biology Corp KCTD16 Antibody - N-terminal region (ARP57447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD16. Alternate names: DKFZp781A1155, KIAA1317, MGC138167ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62133