Learn More
Aviva Systems Biology Corp KCTD18 Antibody - N-terminal region (ARP35421_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35421_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD18. Alternate names: 6530404F10RikApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55727