Learn More
Aviva Systems Biology Corp KCTD21 Antibody - middle region (ARP54467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD21. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human KCTD21. Alternate names: KCASH2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-930