Learn More
Aviva Systems Biology Corp Kctd3 Antibody - middle region (ARP35254_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kctd3. It was validated on Western Blot. . Alternate names: 4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-457