Learn More
Aviva Systems Biology Corp KCTD4 Antibody - N-terminal region (ARP35573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD4. Alternate names: bA321C24.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55782