Learn More
Aviva Systems Biology Corp KCTD7 Antibody - N-terminal region (ARP35422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCTD7. Alternate names: FLJ32069, EPM3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55728