Learn More
Aviva Systems Biology Corp KCTD9 Antibody - middle region (ARP56987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCTD9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human KCTD9. Alternate names: FLJ20038ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-250