Learn More
Organomation 500mL KD Flasks for Central Collection
SDP
Catalog No. NC2030072
Encompass_Preferred
This glassware set contains a 500 mL KD flask, 3-ball Snyder column, 10 mL insulated concentrator tube, and Hopkins condenser for central collection.
Catalog No. NC2030072 Supplier Organomation Supplier No. GS2153C
This glassware set contains a 500 mL KD flask, 3-ball Snyder column, 10 mL insulated concentrator tube, and Hopkins condenser for central collection.