Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KD Scientific Legato™ Series Adapter for Hamilton™ Syringes

Catalog No. 10989022 Shop All BTX Harvard Products
Change view
Click to view available options
For Use With (Equipment):
Gastight Syringe (25 and 50 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-989-022 Gastight Syringe (25 and 50 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-989-022 Supplier KD Scientific Supplier No. 788314
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For 25, 50 and 100 mL Hamilton™ Gastight™ Syringe

  • Selection of accessories and replacement parts for your Legato™ Series Pumps
  • Required for withdraw function

Specifications

Latex Free Yes
Product Type Adapter
Disposable Reusable
For Use With (Equipment) Gastight Syringe (25 and 50 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.