Learn More
KD Scientific Rack for Legato Infusion Withdrawal Pump
Catalog No. 10989007 Shop All BTX Harvard Products
Change view
Click to view available options
For Use With (Equipment):
Legato Series Pumps
Catalog No. For Use With (Equipment)
10-989-007 Legato Series Pumps
1 options
Catalog No. 10-989-007 Supplier KD Scientific Supplier No. 788302
Add to Cart
Add to Cart
Legato 200/201 Plastic/Glass Syringe Microliter Rack
  • Selection of accessories and replacement parts for your Legato™ Series Pumps
  • Accommodates up to four 0.5 μL to 10 mL plastic or glass syringes
Specifications
Holds Plastic or Glass Syringes (Up to four 0.5μL to 10 mL)
Positions 4
For Use With (Equipment) Legato Series Pumps
Product Line Legato
Product Type Microliter Syringe Rack