Learn More
Bioss KD-Validated ABCE1 Rabbit Reco
SDP
Catalog No. 5000066619
Encompass_Preferred
ATP Binding Cassette Subfamily E Member 1 Primary Antibody Unconjugated Public Immunogen Range 460-500/551 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66619 Supplier Bioss Supplier No. BSM90402R
ATP Binding Cassette Subfamily E Member 1 Primary Antibody Unconjugated Public Immunogen Range 460-500/551 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide