Learn More
Bioss KD-Validated ABHD5 Rabbit Reco
SDP
Catalog No. 5000066805
Encompass_Preferred
1-acylglycerol-3-phosphate O-acyltransferase ABHD5 Primary Antibody Unconjugated Public Immunogen Range 180-200/349 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66805 Supplier Bioss Supplier No. BSM90088R
1-acylglycerol-3-phosphate O-acyltransferase ABHD5 Primary Antibody Unconjugated Public Immunogen Range 180-200/349 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide