Learn More
Bioss KD-Validated ACAP2 Rabbit Reco
SDP
Catalog No. 5000065027
Encompass_Preferred
ArfGAP With Coiled-Coil Ankyrin Repeat And PH Domains 2 Primary Antibody Unconjugated Public Immunogen Range 650-720/778 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65027 Supplier Bioss Supplier No. BSM90001R
ArfGAP With Coiled-Coil Ankyrin Repeat And PH Domains 2 Primary Antibody Unconjugated Public Immunogen Range 650-720/778 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide