Learn More
Bioss KD-Validated ACBD3 Rabbit Reco
SDP
Catalog No. 5000065007
Encompass_Preferred
Acyl-CoA Binding Domain Containing 3 Primary Antibody Unconjugated Public Immunogen Range 1-61/528 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65007 Supplier Bioss Supplier No. BSM90092R
Acyl-CoA Binding Domain Containing 3 Primary Antibody Unconjugated Public Immunogen Range 1-61/528 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide