Learn More
Bioss KD-Validated Acetyl-CoA acetyl
SDP
Catalog No. 5000064974
Encompass_Preferred
Acetyl-CoA Acetyltransferase 2 Primary Antibody Unconjugated Public Immunogen Range 200-397/397 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64974 Supplier Bioss Supplier No. BSM90209R
Acetyl-CoA Acetyltransferase 2 Primary Antibody Unconjugated Public Immunogen Range 200-397/397 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide