Learn More
Bioss KD-Validated Acetyl-CoA acyltr
SDP
Catalog No. 5000066880
Encompass_Preferred
Acetyl-CoA Acyltransferase 2 Primary Antibody Unconjugated Public Immunogen Range 360-397/397 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66880 Supplier Bioss Supplier No. BSM90148R
Acetyl-CoA Acyltransferase 2 Primary Antibody Unconjugated Public Immunogen Range 360-397/397 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide