Learn More
Bioss KD-Validated ACSS2 Rabbit Reco
SDP
Catalog No. 5000066943
Encompass_Preferred
Acyl-CoA Synthetase Short Chain Family Member 2 Primary Antibody Unconjugated Public Immunogen Range 100-125/701 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66943 Supplier Bioss Supplier No. BSM90338R
Acyl-CoA Synthetase Short Chain Family Member 2 Primary Antibody Unconjugated Public Immunogen Range 100-125/701 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide